CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20202-48 20202-50 20202-52
MFLX20202-48EA 244 EUR
MFLX20202-48 MFLX20202-50 MFLX20202-52 MFLX20202-51 MFLX20202-49 MFLX20202-53
CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Fittingen en connectoren
Provide easy, secure, sterile connections even in nonsterile environments.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.
All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.