CPC® AseptiQuik-X® Hose Barb Connectors, High Flow
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30003-22 30003-24 30003-25 30003-26 30003-36 30003-31 30003-32
MFLX30003-22EA 241 EUR
MFLX30003-22 MFLX30003-24 MFLX30003-25 MFLX30003-26 MFLX30003-36 MFLX30003-31 MFLX30003-32
CPC® AseptiQuik-X® Hose Barb Connectors, High Flow
Fittingen en connectoren
Provide easy, secure sterile connections even in nonsterile environments.
  • Quick, three-step connection process reduces risk of operator error
  • Large housing size allows easy installation
Manufactured from sturdy polycarbonate, these connectors come presterilized and are for single-use and sanitary applications. Use connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. Push the connectors together; an audible snap confirms a tight connection.More Product Information