LMI Service Kits for Solenoid Diaphragm Metering Pumps
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74513-08 74513-18 74513-38 74513-48 74522-64 74522-68
MFLX74513-08EA 180 EUR
MFLX74513-08 MFLX74513-18 MFLX74513-38 MFLX74513-48 MFLX74522-64 MFLX74522-68
LMI Service Kits for Solenoid Diaphragm Metering Pumps
Pompen Diaphragm Pumps
Optimize performance of your LMI Solenoid Diaphragm Metering Pumps.
  • Keep your pump running smoothly with these service kits
Replace wearable parts as part of a regular maintenance program to prevent failures and downtime.