Accessories for Liquid-Solids Separator / Concentrator Units
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
29995-50 29995-52 29995-54 29995-60
MFLX29995-50EA 79.5 EUR
MFLX29995-50 MFLX29995-52 MFLX29995-54 MFLX29995-60
Accessories for Liquid-Solids Separator / Concentrator Units
Pompen Centrifugal Separators
Optimize performance of your Liquid-Solids Separator.
  • Keep your Separators running properly with these kits
Choose from a wide range of replacement parts and accessories to ensure top performance of your Liquid-Solids Separator / Concentrator Units. Purge kits are available in manual or automatic forms so solids may be purged or bled as needed.