Accessories for Piston Metering Pumps
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX07115-93EA 54.5 EUR
MFLX07115-93 MFLX07115-92 MFLX07115-91
Accessories for Piston Metering Pumps
Pompen Piston Pumps
A wide election of adaptors available to suit FMI Pumps.
  • Constructed of durable PVDF
The integrally moulded port fittings on the standard FMI Type K pump heads accept all 1/4” OD tubing. For other tubing arrangements, special port adapters are required.