Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX33501-16EA 9370 EUR
MFLX33501-16 MFLX33501-17 MFLX33501-36 MFLX33501-73 MFLX33501-82 MFLX33501-15 MFLX33501-24 MFLX33501-25 MFLX33501-91
Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Fluid Sensors
Obtain accurate flow readings without introducing a new material to the fluid path.
  • Noninvasive design eliminates contact with working fluid
  • Ideally suited for strict hygienic standards
  • Continuously monitor peristaltic pump flow rates and send data directly to your computer
  • Compact aluminum sensor body clamps around the tubing
  • Integrated electronics—no external electronic board is needed
  • Specifically calibrated for use with Masterflex L/S® and I/P® platinum-cured silicone tubing
Masterflex® ultrasonic flow sensors accurately and noninvasively measureMore Product Information