GoatThroat™ Spillproof Barrel Pump Accessories
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75650-62 75650-68 75650-92 75650-94
MFLX75650-62EA 66.6 EUR
MFLX75650-62 MFLX75650-68 MFLX75650-92 MFLX75650-94
GoatThroat™ Spillproof Barrel Pump Accessories
Pompen Vatpompen
Enhance versitility of your GoatThroat™ Spillproof Barrel Pump.
  • Optimize performance of your GoatThroat™ Spillproof Barrel Pumps
Accessories for GoatThroat Spillproof Barrel Pump optimize performance or suitable for maintenance.
The pneumatic air adapter, model 75650-40, allows GoatThroat™ pumps to be used with shop air, air compressors, or bottle compressed air. Uses up to 4 psi to deliver fluids up to 4.5 GPM. External pressure valve keeps containers from being over pressurized.