Masterflex® Printed Agglutination Slides, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75955-07 75955-05
MFLX75955-07EA 304 EUR
MFLX75955-07 MFLX75955-05
Masterflex® Printed Agglutination Slides, Avantor®
Object- en dekglaasjes Objectglaasjes
No more makeshift slides.
  • Large wells clearly show reaction results
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Choose numbered/unnumbered—Unnumbered slides allow you to customize the labeling
Use these printed slides to perform latex and other agglutination tests for quick microbe identification. Choose between eight or twelve wells, square or circular, and numbered or unnumbered to suit your application.