Bürkle Cylindrical Barbed Y-Connectors, PVDF
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06044-15 06044-16 06044-17 06044-18 06044-19
MFLX06044-15EA 23.1 EUR
MFLX06044-15 MFLX06044-16 MFLX06044-17 MFLX06044-18 MFLX06044-19
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Fittingen en connectoren
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed Y connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.