Hose Fitting Band Clamps, Stainless Steel
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33350-14 33350-16 33350-18
MFLX33350-12EA 112 EUR
MFLX33350-12 MFLX33350-14 MFLX33350-16 MFLX33350-18
Hose Fitting Band Clamps, Stainless Steel
Klemmen Slangklemmen
Quick and easy assembly without tools.
  • Constructed of durable stainless steel
Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.