Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40614-38
MFLX40614-38EA 14.2 EUR
MFLX40614-38 MFLX40614-40 MFLX40614-41 MFLX40614-42 MFLX40614-43 MFLX40614-44 MFLX40614-45 MFLX40614-46 MFLX40614-47 MFLX40614-48 MFLX40614-49 MFLX40614-50
Masterflex® Union Fittings, Hose Barb, Straight, PVDF, Avantor®
Fittingen en connectoren
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.