Masterflex® Union Fittings, Hose Barb, Straight, Stainless Steel, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30620-01EA 21.4 EUR
MFLX30620-01 MFLX30620-02 MFLX30620-03 MFLX30620-04 MFLX30620-05 MFLX30620-06 MFLX31208-00 MFLX31208-02 MFLX30620-07 MFLX30620-08 MFLX31208-01 MFLX31208-04
Masterflex® Union Fittings, Hose Barb, Straight, Stainless Steel, Avantor®
Fittingen en connectoren
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.