Fittings, Compression Nut Assembly with Ferrule
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31212-63 31212-64 31212-65 31212-66 31212-67 31212-68 31212-69 31212-70 31212-71
MFLX31212-63EA 36.3 EUR
MFLX31212-63 MFLX31212-64 MFLX31212-65 MFLX31212-66 MFLX31212-67 MFLX31212-68 MFLX31212-69 MFLX31212-70 MFLX31212-71
Fittings, Compression Nut Assembly with Ferrule
Fittingen en connectoren
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 220 psi maximum at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).