Union Fittings, Female NPT Threaded, Elbow
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06546-32 06546-33 06546-34 06546-35
MFLX06546-32EA 66.8 EUR
MFLX06546-32 MFLX06546-33 MFLX06546-34 MFLX06546-35
Union Fittings, Female NPT Threaded, Elbow
Fittingen en connectoren
Chemically-inert PFA handles most aggressive chemicals.
  • Quick and easy assembly without tools
Threaded fittings offer a more permanent installation. Choose the size to meet your application needs.