Plug Fittings, Male NPT Threaded, Straight
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
06546-37 06546-38 06546-39 06546-40 34030-55 34030-56 34030-57 34030-58
MFLX06546-37EA 33.2 EUR
MFLX06546-37 MFLX06546-38 MFLX06546-39 MFLX06546-40 MFLX34030-55 MFLX34030-56 MFLX34030-57 MFLX34030-58
Plug Fittings, Male NPT Threaded, Straight
Fittingen en connectoren
Deigned for permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 200 psi at 77 °F ( 25 °C). Usable from -65 to 190 °F (-53 to 87 °C).