Masterflex®-adapterfittingen, vrouwelijke luer naar slangpilaar, schot, polypropyleen, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX45508-30EA 37.4 EUR
MFLX45508-30 MFLX45508-32 MFLX45508-34
Masterflex®-adapterfittingen, vrouwelijke luer naar slangpilaar, schot, polypropyleen, Avantor®
Fittingen en connectoren
Verzeker uw procesintegriteit met volledige materiaaltraceerbaarheid.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging