Label printers EMEA, M211
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
170381 170390
BRDY170381EA 125 EUR
BRDY170381 BRDY170390
Label printers EMEA, M211
Printers Label Printers
Seamlessly design, preview and print labels from your familiar smartphone and swiftly connect to a reliable, industrial grade printer. No other phone-driven label printer today matches the label design possibilities of the M211 Label Printer.
  • Use the M211 Label Printer, Express Labels Mobile App and a phone to create and print labels
  • Tough, industrial labeler designed to survive and thrive on any worksite.
  • Get more done all day long with 300 label prints from one full battery charge
  • Fast and dependable Bluetooth connection
  • Design, preview and print even complex labels with our industry leading Express Labels App
  • Easily create labels using data from your existing spreadsheets
  • Work efficiently by saving and swiftly sharing label designs with colleagues
Design, preview and print – all from your phone. Simple and intuitive, the M211 is built for the job site, running all day on a full charge while resisting drops, shocks and crushes. Never before did an entry level portable printer combine all of these in a single, game-changing device.
Select from 90+ cartridges, filled to the brim with reliable, tested and dedicated label materials that stay attached and remain legible on every surface and in any environment.
Certificaten: Resistant to 1,8 m drops, Resistant to 115 kg crushes, Resistant to military-grade shocks (MIL-STD-810G).
Leveringsinformatie: The printer (only) ships in a carton pack with M21-750-499 label cartridge, AC power cord with EU, UK and US plugs and Quick Start Guide.
The kit ships in a hard case with a M211 printer, M21-750-499 label cartridge, quick start guide, AC power cord with EU, UK and US plugs, belt clip and power pack.