Fragrance allergen mix A2, 40 components in tert-Butyl methyl ether, certified reference material, TraceCERT®, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16558-2ML 16558-5ML
16558-2MLEA 0 EUR
16558-2ML 16558-5ML
Fragrance allergen mix A2, 40 components in tert-Butyl methyl ether, certified reference material, TraceCERT®, Supelco®
Standaarden Organische standaarden
Ready to use certified reference material mixture suitable as calibrant for te testing of allergenic fragrances and their isomers according th the IFRA method.40 analytes in tert-Butyl methyl ether (MTBE).
  • Certification of each individual component by qNMR (following ISO/IEC 17025 accreditation)
  • Mixes produced following an established workflow according to ISO 17034
  • Tested for homogeneity and long-term stability using GC-MS and GC-FID
  • Supplied with a comprehensive certificate including overall uncertainty as well as the individually determined uncertainties of all the components
Components:
3-Propylidenephthalide
α-Acetyl cedrene
Isoeugenyl acetate
α-Amylcinnamaldehyde, predominantly trans
Amyl salicylate
Benzaldehyde
Benzyl benzoate
Benzyl cinnamate
Benzyl salicylate
Butylphenyl methylpropional
D-Camphor
(+)-Carvone
trans-Cinnamaldehyde
Citral total
Coumarin
Damascenone
α-Damascone
(E)-β-Damascone, reference material
β-Damascone
δ-Damascone
Dimethylbenzylcarbinyl acetate (DMBCA acetate)
Eugenyl acetate
Galaxolide 1 (Hexamethylindanopyran)
Galaxolide 2(Hexamethylindanopyran)
Geranyl acetate
Hexadecanolactone / Dihydroambrettolide
α-Hexylcinnamaldehyde
Hydroxycitronellal
Isomethylionone alpha
Linalyl acetate
Hydroxyisohexyl 3-cyclohexene carboxaldehyde
Methyl salicylate
Folione (Methyl 2-octynoate)
Salicylaldehyde
Terpinolene
Vanillin
ISO E alpha + beta+ gamma