Protein standards
Leverancier: Genscript
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M00516 M00521 MM1397-500 M00624-250 M00624-1250
GENSM00516EA 82.6 EUR
GENSM00516 GENSM00521 GENSMM1397-500 GENSM00624-250 GENSM00624-1250
Protein standards
Reagentia voor elektroforese Molecuulgewichtmerkers
GenScript offers a wide selection of Protein molecular weight standards for SDS-PAGE and Western blot, including prestained, unstained, peptide, Western-viewable, and other standards.
  • Comprehensive selection: a variety of standards available for SDS-PAGE and Western blotting
  • Broad molecular weight range: low, middle and broad MW range standards suitable for all practical needs
  • Easy identification: clean, sharp protein bands for accurate MW estimation and assessment of transfer efficiency
The Pre-stained Protein Standards consist of colored bands for easy identification and allow you to monitor the progress of an electrophoretic run. The Unstained Protein Standards provide bands that are unmodified by the presence of dye, for accurate molecular weight estimation of SDS-PAGE. Available in convenient package sizes, all standards have undergone stringent QC to assure consistent performance to provide the sharper bands in the broader molelular weight range available.