Dulbecco's modification of Eagle's medium, MP Biomedicals
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
091033120
091033122
ICNA091033120EA
55.1
EUR
ICNA091033120
ICNA091033122
Dulbecco's modification of Eagle's medium, MP Biomedicals
Celcultuurmedia
Dulbecco's Modified Eagle's Medium (DMEM) with L-Glutamine with 4,5 g/L glucose without sodium bicarbonate is the most widely used modification of Eagle's Basal Medium (BME).
- Ideally for supporting and maintaining a vast range of mammalian cell types
- It contains four times greater concentration of amino acids, vitamins and supplementary components
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures. For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available.