LC-MS/MS system, SCIEX Triple Quad 4500
Leverancier: AB SCIEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5062222 5049829
904-0019EA 256680 EUR
904-0019 904-0091
LC-MS/MS system, SCIEX Triple Quad 4500
Spectrofotometers en fotometers Mass Spectrometers
The Sciex Triple Quad 4500 system is a high sensitivity, bench top triple-quadrupole mass spectrometer designed for LC-MS/MS analyses. This instrument provides excellent robustness and long-term stability for the most demanding assays.
  • The eQ™ Electronics and proven Qurved LINAC® collision cell provide unmatched support for fast chromatography applications with thousands of analytes
  • Turbo V Source: provides high sensitivity analysis over a wide range of flow rates with 'quick change' APCI and TurboIonSpray® probes
  • QJet® ion guide provides better collisional focusing to enhance ion transmission for ultimate sensitivity
  • AcQuRate pulse counting detector enables more accurate and precise ion detection over a wide dynamic range
The 4500 system is a great fit for applications requiring 'round the clock' quantitation, with the robustness to handle repeated injections of complex samples and reliability to minimise downtime. Such applications include food allergen testing, testing 93 forensic drugs in urine, amino acid quantitation, and gluten quantitation in foods.
Certificaten: NRTL certificate and EU Declaration of Conformity.
Informatie over toebehoren: TQ/QT software starter kit bundle contains Analyst 1.7 Acquisition and Processing License for acquisition and processing on a single workstation, Analyst 1.7 Processing License for processing only (this license does not allow acquisition control), and Sciex OS-MQ 2.1 license for quantitation of small molecule compounds, large molecule biomarkers and biopharmaceuticals on SCIEX LC-MS systems.
DiscoveryQuant software version 3.0 supports HT-ADME in the drug discovery laboratory. Combined 'Optimise' and 'Analyse' functionality optimises MS analytical conditions and builds high throughput discovery assays quickly and efficiently from a single view. Package includes a single-instrument license that activates 'Analyse' and 'Optimise' portions of the software. Supports most SCIEX Triple Quad, QTRAP And TripleToF platforms, with exception of Triple Quad 3500 systems. One license, no software DVD, SW downloadable from the SCIEX website. Compatible with Windows 7 and 10 operating systems.
MS Office 2016 Professional plus ISV global kit - Microsoft Office Professional Plus 2016 w/SP1. This is an English version of Microsoft Office that can be shipped and activated in all global regions. This kit contains a disc and license key that can only be activated by SCIEX personnel (purchase of the above listed product is governed by the terms and conditions found at https://www.microsoft.com/en-us/licensing/product-licensing/products.aspx).
SCIEX OS-Q 2.1; Qual and quant processing perpetual license - SCIEX OS-Q 2.1 perpetual license for processing data on an additional PC. SCIEX OS is a leap forward in data processing analytics. The all new interface is much easier to use and learn while incorporating the power of a quantitative and qualitative evaluation of data in one step. This license is for processing only, data acquisition requires an acquisition license. Software can be downloaded or a DVD purchased separately.
SCIEX OS-MQ 2.1; Quant only processing perpetual license - SCIEX OS-MQ 2.1 perpetual license for processing data on an additional PC. SCIEX OS is a leap forward in data processing analytics. The all new interface is much easier to use and learn while incorporating the power of a quantitative evaluation of data in one step. This license is for processing only, data acquisition requires an acquisition license. Software can be downloaded or a DVD purchased separately.
SCIEX OS 2.1; 21 CFR Part 11 Compliance ready perpetual e-license - SCIEX OS 2.1 perpetual e-License for 21 CFR Part 11 compliance tools. This 21 CFR Part 11 compliance ready add-on, allows the user to enable e-signatures and audit trails to allow them to meet 21 CFR Part 11 guidelines. This is an add-on that requires an acquisition or processing license to function. Software can be downloaded or a DVD purchased separately.
PEAK MS bench G SCI 1 - gas generator enclosure bench, integrated with model Genius 1024. Built-in air compressor, no external air source required. Maximum gas flows of curtain (N₂) 19 L/min, source (air) 26 L/min, and exhaust (air) 25 L/min. Bench top ideally suited as space for Liquid Chromatography (LC) system. Bench includes soundproof panels. Designed for use with any SCIEX RUO system. Dimensions (W×D×H): 91,4×78,7×76,2 cm (36×31×30"), weight 120 kg (265 lbs). Electrical requirements 220 to 240 V, 50/60 Hz, 7,0 A.
PEAK MS bench SCI 1 - MS instrument bench with room for vacuum pumps within enclosure. Includes soundproof panels. Reduces noise and vibration of roughing pumps while providing fan-assisted cooling. Dimensions (W×D×H): 91,4×78,7×76,2 cm (36×31×30"), weight 85 kg (187 lbs). Electrical requirements 100 to 240 V, 50/60 Hz, 2,0 A. Bench supports SCIEX RUO bench top instruments including 5500+, X500 series QTOF, 6500+ series, 6500 series, 5500 series, 4500 series, API3200 series, and Triple Quad 3500 (note: This part number does not include an integrated gas generator).
MS bench - instrument bench suitable for all SCIEX bench top MS systems. Features holes for hose and cable management. Dimensions (W×D×H): 60×31×30".
LC bench - features holes for hose and cable management as well as a removable tray for solvent storage. Dimensions (W×D×H): 36×31×30".
ExionLC 2.0 system component-based binary UHPLC system rated at 860 bar (12473 psi). Includes the following ExionLC 2.0 components: ExionLC 2.0 binary pump with integrated 4-channel degasser and 2-channel solvent selection valve for selection of 4 solvents, ExionLC 2.0 autosampler, ExionLC 2.0 column oven, ExionLC 2.0 solvent tray, ExionLC 2.0 bottle set with SecurityCap system, ExionLC 2.0 waste management system, SecurityLINK SS 100 µm ID× 500 mm (qty 2), and accessories. Recommended configuration for routine HPLC/UHPLC applications. Requires 4 power cords, sold separately.
ExionLC 2.0+ system component-based high pressure binary UHPLC system rated at 1240 bar (17985 psi). Includes the following ExionLC 2.0+ system components: ExionLC 2.0 binary pump+ with integrated 4-channel degasser and 2-channel solvent selection valve for selection of 4 solvents, ExionLC 2.0 autosampler+, ExionLC 2.0 column oven, ExionLC 2.0 solvent tray, ExionLC 2.0 bottle set with SecurityCap system, ExionLC 2.0 waste management system, SecurityLINK SS 100 µm ID× 500 mm (qty 2), and accessories. Recommended configuration for high pressure UHPLC LC-MS applications. Requires 4 power cords, sold separately.
ExionLC 2.0 Quaternary system component-based quaternary UHPLC system rated at 1240 bar (17985 psi). Includes the following ExionLC 2.0 system components: ExionLC 2.0 LPG pump, ExionLC 2.0 autosampler, ExionLC 2.0 column oven, ExionLC 2.0 solvent tray, ExionLC 2.0 bottle set with SecurityCap system, ExionLC 2.0 waste management system, SecurityLINK SS 100 µm ID× 500 mm (qty 2), and accessories. Requires 4 power cords, sold separately.
ExionLC 2.0 binary pump - ExionLC 2.0 binary pump rated to 860 bar (12473 psi), ideal for routine HPLC/UHPLC applications. Includes integrated 4-channel degasser, 2-channel solvent selection valve for selection of 4 solvents, 50 µl mixer. Requires power cord and accessories, sold separately.
ExionLC 2.0 binary pump+ - ExionLC 2.0 binary pump rated to 1240 bar (17985 psi), ideal for more demanding UHPLC applications. Includes integrated 4-channel degasser, 2-channel solvent selection valve for selection of 4 solvents, 50 µl mixer. Requires power cord and accessories, sold separately.
ExionLC 2.0 LPG pump - ExionLC 2.0 low pressure gradient pump rated to 860 bar (12473 psi) for blending up to 4 solvents for quaternary gradient formation. Includes integrated 4-channel degasser, and 100 µl mixer. Requires power cord and accessories, sold separately.
ExionLC 2.0 autosampler - ExionLC 2.0 temperature controlled autosampler rated to 860 bar (12473 psi). Sample capacity of 96×1,5 ml vials or 2× microtiter plates. Includes 100 µl loop, 2×48-position vial trays. Requires power cord and accessories, sold separately.
ExionLC 2.0 autosampler+ - ExionLC 2.0 temperature controlled autosampler rated to 1240 bar (17985 psi). Sample capacity of 96×1,5 ml vials or 2× microtiter plates. Includes 100 µl loop, 2x48-position vial trays. Requires power cord and accessories, sold separately.
ExionLC 2.0 wash system - high performance autosampler wash system for enhanced carryover management with the flexibility to select from up to 8 wash solvents with delivery rates of up to 4 ml/min. Includes active needle wash feature. Tubing and fittings for 4 wash solvents standard, wash system expansion kit required to support 8 solvents. Requires power cord, sold separately.
ExionLC 2.0 wash system expansion kit - wash system expansion kit includes accessories to extend standard wash system capacity by an additional 4 wash solvents for the selection of a total of 8 wash solvents.
ExionLC 2.0 column oven - ExionLC 2.0 column oven with Peltier temperature control from 5 °C below ambient temperature to 85 °C. Accommodates up to 4 columns of 30 cm length or 8 columns of 15 cm length. Includes solvent pre-heater. Requires power cord and accessories, sold separately.
Kit, ExionLC 2.0 diode array detector - ExionLC 2.0 diode array detector with deuterium lamp for a wavelength range of 190 to 700 nm. With advanced optical unit design and temperature management for maximum sensitivity and minimal baseline drift. Includes ExionLC 2.0 detector flow cell 50 bar and 1 SecurityLINK SS 100µm ID× 500 mm. Requires power cord, sold separately.
Kit, ExionLC 2.0 diode array detector HS
Kit, ExionLC 2.0 multi-wavelength detector - ExionLC 2.0 multi-channel UV detector with deuterium lamp for a wavelength range of 190 to 700 nm. Includes ExionLC 2.0 detector flow cell 300 bar and 1 SecurityLINK SS 100 µm ID× 500 mm. Requires power cord, sold separately.
ExionLC 2.0 diode array detector - ExionLC 2.0 diode array detector with deuterium lamp for a wavelength range of 190 to 700 nm. With advanced optical unit design and temperature management for maximum sensitivity and minimal baseline drift. Accessories sold separately.
ExionLC 2.0 diode array detector HS - ExionLC 2.0 high sensitivity UV/Visible diode array detector with deuterium and tungsten halogen lamps for a wavelength range of 190 to 1000 nm. With advanced optical unit design and temperature management for maximum sensitivity and minimal baseline drift. Accessories sold separately.
ExionLC 2.0 multi-wavelength detector - ExionLC 2.0 multi-channel UV detector with deuterium lamp for a wavelength range of 190 to 700 nm. Accessories sold separately.
Bestelinformatie: Standard parts and labour warranty for 1 year starting from the completion of instrument commissioning. Upgradeable to QTRAP functionality with article ABSX5049829.