Wire baskets, round, with handle
Leverancier: RSG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
80120080 80120180
216-2376EA 57.4 EUR
216-2376 216-2377
Wire baskets, round, with handle
Manden Draadmanden
Made from stainless steel 1.4301.
  • Rust-proof
  • Very good temperature resistance
  • Good chemical resistance
  • With reinforced edges