Multiple affinity removal column human 14
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5188-6557 5188-6558
AGLS5188-6557EA 5457 EUR
AGLS5188-6557 AGLS5188-6558
Multiple affinity removal column human 14
Kolommen voor chromatografie
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
  • Made of PEEK (polyetheretherketone)
  • High-capacity serum load
  • Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.