Petri Dishes, Glass, Assistent®
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
41042030 41042006 41042024 41042036
HECH41042030EA 216 EUR
HECH41042030 HECH41042006 HECH41042024 HECH41042036
Petri Dishes, Glass, Assistent®
Cultuurschalen Petrischalen
Soda-lime glass, with lid. Especially for use as disposables.
  • Good chemical resistance
  • Heatable up to +60 °C
  • Not suitable for applications related to strong heat or rapid temperature changes
  • Ideal for bacterial culture production
  • Single compartment
  • No vents