Dimethylsulfoxide ≥99.9%, Cryopres™ USP for cryopreservation, sterile-filtered
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
092780145 092780148
092780145.EA 42.3 EUR
092780145. 092780148.
Dimethylsulfoxide ≥99.9%, Cryopres™ USP for cryopreservation, sterile-filtered
Dimethylsulfoxide
Cryopres™ Dimethyl sulfoxide is commonly used in cell cryopreservation, along with glycerol, to prevent damage to cell membranes during freezing. USP, Ph. Eur., sterile-filtered.
Opgelet: For laboratory use. Not for injection.
Formule: (CH₃)₂SO
MW: 78.14 g/mol
Kookpunt: 189 °C (literatuur)
Smeltpunt: 16…19 °C (literatuur)
Dichtheid: 1.104 g/cm³ (20 °C)
Vlampunt: 87 °C (188.6 °F) (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002089
CAS nummer: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
Order Now
Specificaties
Environmentally Preferable