CF® Dye cAMP
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00037 00036
00037.EA 204 EUR
00037. 00036.
CF® Dye cAMP
Koppelings- en labelingssystemen voor eiwitten
Fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.
  • Superior CF® dyes are bright, photostable and water-soluble
  • Available with green or far-red labels
  • Soluble in H₂O, DMF, or DMSO
CF®488A-cAMP and CF®640R-cAMP are fluorescently labelled cAMP analogs that can be used to probe cAMP receptors.
Biotium’s next-generation CF® Dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
CF is a registered trademark of Biotium, Inc. Alexa Fluor, Texas Red, and DyLight are registered trademarks of Thermo Fisher Scientific.