STC1 series steam through connector, quick disconnect coupling for bio and pharma
Leverancier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STC1700000 STC1700100 STC1700200 STC1700300 STC1700500 STC1700600 STC1700700 STC1700800
212-0841EA 939 EUR
212-0841 212-0842 212-0843 212-0844 212-0845 212-0846 212-0847 212-0848
STC1 series steam through connector, quick disconnect coupling for bio and pharma
Fittingen en connectoren
Polysulphone connector designed to easily connect stainless steel pharmaceutical process equipment to single use disposable plastic tube sets and media bags.
Materials of construction include USP class 6 animal free (BSE/TSE) polysulphone, USP class 6 andimal free (BSE/TSE) silicone orings and seals.
Connect a bioreactor, media bag and steam trap in one easy, sterile connection.
Certificaten: USP Class VI, BSE/TSE free materials (ADCF)
Bestelinformatie: Accessories not included