MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
Leverancier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MPX17803 MPX17803B25 MPX17839 MPX22803M MPX22839M MPXK17803 MPXK17803B25 MPXK17839
212-0833EA 1600 EUR
212-0833 212-0834 212-0835 212-0836 212-0837 212-0838 212-0839 212-0840
MPX series ¹/₂" flow, quick disconnect coupling for bio and pharma
Fittingen en connectoren
These ¹/₂" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets.
Materials of construction include animal free (BSE/TSE) USP class 6 polysulphone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certificaten: USP Class VI, BSE/TSE free materials (ADCF)
Bestelinformatie: Accessories not included