AseptiQuik® X connectors
Leverancier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AQX33024HT AQX44024HT
CPCCAQX33024HTEA 231 EUR
CPCCAQX33024HT CPCCAQX44024HT
AseptiQuik® X connectors
Fittingen en connectoren
AseptiQuik® X large format 1" connectors provide quick and easy sterile connections for high flow applications, even in non-sterile environments. The 'Twist-Pull-Twist' design enables users to quickly transfer large volumes of media easily with less risk of operator error.
  • Operating temperature: 4 to 40 °C
  • Ensure simultaneous and secure removal of both membranes
  • Audible confirmation of completed assembly steps
  • Open access through multiple supply chain partners
  • Secures final connection preventing disassembly
Biopharmaceutical manufacturers can now make 1" sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Certificaten: USP Class VI, ADCF Certified.