Beaker tongs, stainless steel
Leverancier: RSG Solingen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RSGA80.043.330EA 35.6 EUR
RSGA80.043.330
Beaker tongs, stainless steel
Tangen
Beaker/cup tongs are made from stainless steel 1.4301.
  • Electrolytically polished
  • Jaws wrapped with organic fleece