V Spatula sampling tools
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1095N-150 1095N-250
300-0426EA 111 EUR
300-0426 300-0427
V Spatula sampling tools
Monsternemers Monsternemers voor vaste stoffen
Ideal for taking surface samples of powders and viscous materials.
  • Multi-purpose sampling tool, available in a range of models
  • Suitable for sampling cheeses, waxes, and other hard materials
  • Robust V shaped blade allows for easy emptying
  • Made from chemically resistant stainless steel 316L
The V Spatula is available in varying models to offer the right solution. It is ideal for taking surface samples of a wide variety of substances, such as powders and harder materials such as waxes and cheeses, thanks to its robust blade design.