Trolley for collecting trays, b.safe
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R510-05 R510-06 R510-10 R510-11
BOHLR510-05EA 312 EUR
BOHLR510-05 BOHLR510-06 BOHLR510-10 BOHLR510-11
Trolley for collecting trays, b.safe
Transportwagens
Made of aluminium profiles and polyamide connectors.
  • With four casters
  • Two casters with brakes
  • Flexibly placed and transported containers
Canisters can be flexibly placed under the work bench. Full containers can be transported.
Order Now
Specificaties
Environmentally Preferable