L-Phenylalanine 98,5-101,5 Dried USP, multi-compendial, Macron Fine Chemicals™
Leverancier: Avantor Performance Materials
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
M.O.C. Category R
MACR4945-06EA 1546.31 EUR
MACR4945-06 MACR4945-07
L-Phenylalanine 98,5-101,5 Dried USP, multi-compendial, Macron Fine Chemicals™
L-Phenylalanine
Formule: C₉H₁₁NO₂
MW: 165.19 g/mol
Kookpunt: 313 °C (1013 hPa)
Smeltpunt: 173 °C
Dichtheid: 0.754 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064227
CAS nummer: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271