Drinking straws
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
44030015 44030020 44030023 44030115 44030120 44030123
HECH44030015EA 220 EUR
HECH44030015 HECH44030020 HECH44030023 HECH44030115 HECH44030120 HECH44030123
Drinking straws
Foodservice benodigdheden
Reusable drinking straws are made from borosilicate glass 3.3.
  • Straight form, with fused edges
  • Outer diameter: 9 mm
  • Inner diameter: 6 mm
  • Autoclavable
Ideal for use in hospitals, these straws are highly resistant against chemicals and thermal shock.