Upright microscopes, Leica DM4 B and DM6 B
Leverancier: LEICA MICROSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11888857_P1 11888866_P1
630-3466EA 0 EUR
630-3466 630-3467
Upright microscopes, Leica DM4 B and DM6 B
Microscopen
Increase your work efficiency with the Leica DM4 B or Leica DM6 B upright digital research microscopes. Both are ideally suited to making your work life easier in biomedical research and clinical labs alike.
  • Simplify your workflow with automated functions and easy-to-use software
  • Easily capture publication-quality images by utilising the 19 mm sCMOS camera imaging port
  • Create fast overviews of your samples and identify the important details instantly with the LAS X navigator software
  • Have great flexibility through the choice of accessories
  • Change contrast with the push of a button – everything you need automatically slides into place
  • Acquire publication-ready images
Focus on the result you would like to achieve rather than the process of getting to it. Intelligent automation does the work for you. It solves multi-step routines with the press of a button, makes your work life easier, saves time, and allows you to focus on your experiment, not on complex microscope settings.
You can configure the microscope that fits your need and budget with the Leica DM4 B and Leica DM6 B. Optimise your system to your application with different contrast methods and automated functions.
LED illumination lights your sample homogenously and with a constant colur temperature, while also saving energy and avoiding bulb changes due to the LED’s lifetime of up to 25000 hours.