Boiling water baths, Hydro series
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L003066 L003067 L003068 L003069 L003070
462-0619EA 933 EUR
462-0619 462-0620 462-0621 462-0622 462-0623
Boiling water baths, Hydro series
Baden Kookbaden
Vaporisation water baths which are ideal for gentle steaming work. The working temperature range is 25 to 100 °C.
  • Temperature adjustment via rotary knob with temperature scale
  • 1-point temperature controller
  • Low water level protection
  • Bath interior, cover, sieve plate and heating element are made of stainless steel
  • All hole covers consist of a multi-part ring set and are removable
  • Multi-piece ring set made of heat-resistant plastic
The cover frame includes an integral hole designed to accommodate a rotor rod.
Temperature stability: ±3 K
Electrical: 230 V; 50/60 Hz
Leveringsinformatie: Supplied with adjustable water level regulator, mesh plate and ring set as standard.