Sample splitters
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SAMP8600M-30EA 758 EUR
SAMP8600M-30 SAMP8600M-20 SAMP8600M-40
Sample splitters
Monsterverdelers
Perfect for reducing a bulk sample into two equal quantities.
  • Includes 2 receptacles for collecting the sample
  • Ideal for food, environmental and chemical use
  • Easy to use
The Steel Sample riffle boxes (splitters) removes the difficulty from the important task of dividing or reducing a bulk sample into two equal quantities. Each riffler comes with 2 receptacles for collecting the sample after it has gone through the splitter. The rifflers are ideal for food, environmental and chemical use. They are very easy to use. Simply drop the sample into the top and the sample is split into two. The split sample drops into the riffle boxes underneath. We recommend that that the width of the slots are 2.5 to 3 times bigger than the diameter of the largest particle being split.