Stainless Steel Buckets
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A586-10 A586-12 A586-15
SAMPA586-10EA 368 EUR
SAMPA586-10 SAMPA586-12 SAMPA586-15
Stainless Steel Buckets
Emmers
These general purpose buckets are ideal for use in all areas of industry.
  • Made from chemically resistant 316L stainless steel
  • Strong and robust heavy duty design
  • Perfect for corrosive environments
The range of stainless steel buckets from Sampling Systems are excellent multi purpose buckets that can be used across a wide range of industries. Made from chemically resistant 316L stainless, they are particularly suitable for use in corrosive environments.