Masterflex® 1-1/2” Sanitary Tri-Clamp Cap, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX40330-19EA 289 EUR
MFLX40330-19 MFLX40330-20 MFLX40330-21
Masterflex® 1-1/2” Sanitary Tri-Clamp Cap, Avantor®
Fittingen en connectoren
Close off your sanitary fluid line with a secure connection.
  • Fully compatible with other similar sized Tri-Clamp® connections
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Sterilizable via gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. These adapters offer good chemical resistance and may be sterilized by gamma irradiation. Clamps and gaskets required for a complete system are sold separately.