Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX40332-00EA 43.2 EUR
MFLX40332-00 MFLX40332-01 MFLX40332-02 MFLX40332-03 MFLX40332-04 MFLX40332-05 MFLX40332-06 MFLX40332-07 MFLX40332-08 MFLX40332-09
Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Fittingen en connectoren
Ribbed base prevents suction blockage and increases rigidity.
  • Barb sizes range from 1/8" to 1"
  • 1-3/4" and 2-1/4" ribbed bases
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.