Calibration thermostats, ECO
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L002847 L002848 L002850
LAUDL002847EA 9780 EUR
LAUDL002847 LAUDL002848 LAUDL002850
Calibration thermostats, ECO
Circulatiethermostaten Open circulatiesystemen Kalibratiethermostaten
Calibration thermostatic bath with latest microprocessor technology and integrated cooling system. Working temperature range: −25 to 200 °C
  • Safety class III for operation with flammable and non-flammable liquids. Over-temperature cut-out adjustable via menu
  • Various pump with six adjustable performance levels
  • Easy control of the flow rate between internal and external circulation during operation without contact with the bath
  • USB interface as standard
  • 2-chamber construction for constant level in the thermosetting chamber
  • Vertical adjustment of the overflow height of the thermosetting chamber
  • Bath tank made from stainless steel with drain valve
  • Condenser cooling air
  • Utilises natural refrigerants
The calibration thermostat has a coloured TFT display for simultaneous indication of actual and set values and graphic illustration of the temperature profile. User friendly menu navigation in plain language with an easy input function via cursor and soft keys. Additional Tmax key for overtemperature protection and a fully electronic continuous controller with PID action. USB interface as standard, upgradeable with an interface module (analogue module, contact module, RS232/485 module, Profibus, Ethernet-USB module).
Upgradeable with PT100/LiBus module for external control and remote control via Command console.
Bestelinformatie: Calibration racks are available to purchase separately.
Leveringsinformatie: Supplied with nipples, screw caps and bath cover.