High performance centrifuge, Avanti™ J-E
Leverancier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Avanti™
521-4062EA 37059 EUR
521-4062
High performance centrifuge, Avanti™ J-E
Centrifuge Vloercentrifuges
A high-performing centrifuge for tight laboratory spaces. The Avanti J-E centrifuge generates maximum speeds up to 2.1×10⁴ min⁻¹ (53300 ×g) and offers the capacity of processing up-to 4 litres in under 10 minutes. The Avanti J-E's high torque switched reluctance (SR) direct drive achieves faster acceleration and deceleration with gentle precision that delivers the fastest separations in the shortest run times, performance unattainable by standard high-speed centrifuges. The comprehensive standard and BioSafe-certified J-series rotors and labware are all compatible with the J-E, for sample sizes from a few microliters up-to 4 litres.
  • Make the most out of your workspace and workflow
  • Achieve high performance and up to 4,0 L processing capacity in a compact foot print
  • Speeds up to 53300 ×g RCF with efficient acceleration and deceleration profiles
  • The Avanti JE offers an intuitive, user-friendly interface, as well as lower energy costs
  • Heat output: 6900 BTU/hr (2,0 kW)
  • Refrigeration: Non-CFC, non-ozone depleting refrigerant
For total versatility, select the All Spin JS-5.3 high capacity swinging bucket rotor system, which processes up-to twenty-four 96-well microtiter plates and offers the flexibility of processing a variety of sample sizes in bottles from 4×500 ml to 96×17 mm tubes. Using CFC-free refrigerants and a maintenance-free vacuum pump to partially remove friction-generating air molecules, the J-E offers better temperature control (−10 to +40 °C in 1 °C increments), a more energy efficient system, lower heat output (<6900 BTUs) and quieter operation for a comfortable, productive work environment.
Please see High Performance Centrifuge Catalogue section in the PDF catalogue under Documentation for full technical specification.