Zero pressure weather caps
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7095100 5622100 4722200
LBCP7095100EA 0 EUR
LBCP7095100 LBCP5622100 LBCP4722200
Zero pressure weather caps
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.