ESD floor mattings, Notrax® 457 Skystep™
Leverancier: JUSTRITE MANUFACTURING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
457S0023BL 457S0034BL 457S0035BL
115-2864EA 538 EUR
115-2864 115-2865 115-2866
ESD floor mattings, Notrax® 457 Skystep™
Vloermatten Anti-vermoeidheidsmatten
The Skystep™ ESD static dissipative mat is a stand-alone rubber mat designed for individual work stations. Ergonomically engineered, the SkyStep™ ESD utilizes the power of air.
  • Superb anti-fatigue properties due to the ergonomic bubble pattern
  • Rubber molded, air-pocket underside anti-fatigue mat
  • Made of heavy duty natural rubber compound
  • ESD Static Dissipative, measured resistance 105 to 107 Ω
  • Beveled edges on all four sides
  • Grounding cord available
The mat is designed to suction to the ground to prevent shifting while trapping air in its chambers. Combined with the resiliency of the rubber, these air pockets create an ultimate cushioning effect while keeping the mat lightweight. Molded, beveled edges on all four sides prevent trip hazards.