Adjustable volume ergonomic pipettes, single and multichannel, xPIPETTE™
Leverancier: BIOTIX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30459890 30459891 30459892 30459893 30459894 30551495 30459895 30537396 30537397 30537449 30537450 30537451 30537452 30537454 30537455
613-6897EA 656 EUR
613-6897 613-6898 613-6899 613-6900 613-6901 613-6902 613-6903 613-6904 613-6905 613-6906 613-6907 613-6908 613-6909 613-6910 613-6911
Adjustable volume ergonomic pipettes, single and multichannel, xPIPETTE™
Pipetten Micropipetten
xPIPETTES™ offer accuracy and precision with low ejection forces. Biotix uses patented tip ejection technology to reduce fatigue and makes the xPIPETTE a top choice to improve your lab’s ergonomics.
  • Fully compatible with Rainin LTS tips and Biotix xTIP4™ tips
  • Calibrated to ISO 8655 standards with Biotix xTIP4 tips
  • Light spring forces produce a smooth and controlled aspiration with perfect balance
  • Padded ejector button to reduce hand vibrations during tip ejection
  • Simple volume adjustment with a secure lock to prevent volume drift
  • Available in single, 8, and 12-channel models with a symmetrical design for ambidextrous use
These pipettes are designed with durable and lightweight components. They deliver a precise and accurate liquid transfer while maintaining optimal hand comfort.
Certificaten: ISO 8655 certified. Calibration certificate included in the box with each pipette. Unique serial ID number under the finger hook for GLP/GMP compliance.
Opgelet: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.