Darifenacin Peak ID Mixtures, Reference Standards
Leverancier: USP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1164211 1164222
1164211.EA 597 EUR
1164211. 1164222.
Darifenacin Peak ID Mixtures, Reference Standards
Standaarden Organische standaarden
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Darifenacin Peak ID Mixture: A mixture consisting of Darifenacin Hydrobromide (99%), Dealkyl darifenacin (0.3%), Ketodarifenacin (0.2%), and 7-Bromo darifenacin (0.3%)
See USP Certificate for full Chemical Name
Certificaten: US Pharmacopea Reference Standard