Pierce™ Deuterium-labelled Cross Linkers
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21595 21590 21610
PIER21595EA 363 EUR
PIER21595 PIER21590 PIER21610
Pierce™ Deuterium-labelled Cross Linkers
Koppelings- en labelingssystemen voor eiwitten
Cross linkers are used for the identification of near-neighbor protein relationships and ligand-receptor interactions.
  • Deuterium-labelled cross linkers
Deuterium-labelled cross linkers are specially manufactured for crosslinking protein structure: Function analysis by mass spectrometry.