Flexible duct connections
Leverancier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4861800 5621400
LBCP4861800EA 0 EUR
LBCP4861800 LBCP5621400
Flexible duct connections
Ducting Supplies
Flexible duct connections reduce vibration between the blower and PVC ductwork.
Supplied with two clamps for easy installation.