Generators, Polytron® PT-DA/BEC (BIOTRONA)
Leverancier: KINEMATICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11030291 11030286 11030034
KINE11030291EA 2490 EUR
KINE11030291 KINE11030286 KINE11030034
Generators, Polytron® PT-DA/BEC (BIOTRONA)
Homogenisatoren Dispergeerstaven
BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316 L, electropolished, Ra≤1,6 μm as standard.
  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces