HYDRANAL™ - NEXTGEN Coulomat C-FA, Catholyte for Coulometric Karl Fischer Titration, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34470-50ML 34470-6X50ML
HONC34470-50MLEA 424 EUR
HONC34470-50ML HONC34470-6X50ML
HYDRANAL™ - NEXTGEN Coulomat C-FA, Catholyte for Coulometric Karl Fischer Titration, Fluka™
Karl Fischer titranten en reagentia
Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances)
  • Assay of water Max. 30 ppm
  • Number of samples Min. 3